tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
: Merck
: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
:1634-04-4
:
:(CH₃)₃COCH₃
:MFCD00008812
:88.15 g/mol
:13,06059
:55.2 °C (1013 hPa)
:–108.6 °C
:0.7405 g/cm³ (20 °C)
:216-653-1
:2398
:3,II
:M-Clarity™ quality level = MQ100
:–28 °C
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
Frequently Bought Together