Order Entry
Austria
ContactUsLinkComponent
tert-Butyl methyl ether 99%, pure
tert-Butyl methyl ether 99%, pure
Certificates
undefined
tert-Butyl methyl ether 99%, pure
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3675 to 1.3695 (20°C, 589 nm)
  • Water:
    ≤0.01 % (Coulometric)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together