Order Entry
Austria
ContactUsLinkComponent
L(-)-Glutathione (reduced form)
L(-)-Glutathione (reduced form)
Catalog # CAYM703316-1
Supplier:  Cayman Chemical
CAS Number:  
L(-)-Glutathione (reduced form)
Catalog # CAYM703316-1
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :307.33 g/mol
Merck Index:13,04488
Melting Pt:185…195 °C
Density:1.475 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-725-4
CAS Number:70-18-8
Formula:C₁₀H₁₇N₃O₆S
MDL Number:MFCD00065939