Order Entry
Austria
ContactUsLinkComponent
4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Irritant
Flammable
4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 293261-1L
   Safer Human and Environmental Health
CAS Number:  
undefined
Irritant
Flammable
4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 293261-1L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.50 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 380 nm:
    Max. 0.02
  • Absorbance at 360 nm:
    Max. 0.15
  • Absorbance at 340 nm:
    Max. 0.50
  • Absorbance at 335 nm:
    Max. 1.0
  • Explosion Limit:
    1.2-8 %, 93 °F
  • Vapor Density:
    3.5 (vs air)

Specification Test Results

About this item

 :
 :100.16 g/mol
Merck Index:13,05227
Boiling Pt:115-117 °C (1.013 hPa)
Boiling Pt:115…117 °C (1.013 hPa)
Melting Pt:-83 °C
Melting Pt:–83 °C
Density:0.8 g/cm³ (20 °C)
Flash Pt:14 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
 :GHS07
 :GHS02
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938