2-Chloro-2-methylpropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: tert-Butyl chloride
CAS Number:507-20-0
:
Formula:C₄H₉Cl
MDL Number:MFCD00000816
:92.57 g/mol
Merck Index:13,01561
Boiling Pt:51 °C (1013 hPa)
Melting Pt:–28 °C
Density:0.85 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:208-066-4
UN:1127
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–27 °C
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together