Viscosity reference standards
Supplier: PerkinElmer
Jump to:
About this item
8 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- TypePkAvailabilityPrice
- PERCN5316000Viscosity standard reference kit, 5×500 ml
- PERCN9307740Nominal viscosity: 4,5 cSt at 40 °C and 1,6 cSt at 100 °C, 500 mlNominal viscosity: 4,5 cSt at 40 °C and 1,6 cSt at 100 °C, 500 mlHydrocarbons1Log in for availability1 * 1 ST169,00 €
- PERCN5316001Nominal viscosity: 10 cSt at 40 °C and 2,7 cSt at 100 °C, 500 mlNominal viscosity: 10 cSt at 40 °C and 2,7 cSt at 100 °C, 500 mlHydrocarbons1Log in for availability1 * 1 ST169,00 €
- PERCN5316002Nominal viscosity: 30 cSt at 40 °C and 5,3 cSt at 100 °C, 500 mlNominal viscosity: 30 cSt at 40 °C and 5,3 cSt at 100 °C, 500 mlHydrocarbons1Log in for availability1 * 1 ST169,00 €
- PERCN5316003Nominal viscosity: 100 cSt at 40 °C and 16,8 cSt at 100 °C, 500 mlNominal viscosity: 100 cSt at 40 °C and 16,8 cSt at 100 °C, 500 mlHydrocarbons1Log in for availability1 * 1 ST169,00 €
- PERCN5316004Nominal viscosity: 360 cSt at 40 °C and 42 cSt at 100 °C, 500 mlNominal viscosity: 360 cSt at 40 °C and 42 cSt at 100 °C, 500 mlHydrocarbons1Log in for availability1 * 1 ST169,00 €
- PERCN5316005Nominal viscosity: 930 cSt at 40 °C and 82 cSt at 100 °C, 500 mlNominal viscosity: 930 cSt at 40 °C and 82 cSt at 100 °C, 500 mlHydrocarbons1Log in for availability1 * 1 ST169,00 €
- PERCN5316025Nominal viscosity: 30 cSt at 40 °C and 5,3 cSt at 100 °C, 3,8 lNominal viscosity: 30 cSt at 40 °C and 5,3 cSt at 100 °C, 3,8 lHydrocarbons1Log in for availability1 * 1 ST1 030,00 €