ContactUsLinkComponent
J.T.Baker® BAKERBOND Speedisk® Amino (NH2), SPE Columns
J.T.Baker® BAKERBOND Speedisk® Amino (NH2), SPE Columns
Catalog # 8165-03
Supplier:  Avantor
J.T.Baker® BAKERBOND Speedisk® Amino (NH2), SPE Columns
Catalog # 8165-03
Supplier:  Avantor
Specifications
  • Capacity:
    3 ml
  • Bed weight:
    35 mg
  • Phase:
    Amino (NH₂)
  • Pk:
    1
Specifications
About this item
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate prefiltration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Lipids (fatty acids, cholesterol).
These SPE columns can be used in ion exchange and normal phase mode.