



n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis
Catalog # 83962.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.0 %
- Acidity:Max. 0.0005 meq/g
- Evaporation residue (100°C):Max. 0.0005 %
- Water:Max. 0.01 %
- Organic residue (as Octanol):Max. 10 ng/ml
- Halogenated residue (as Lindane)(GC/ECD):Max. 5 ng/l
Specification Test Results
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94,3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119480412-44
:GHS02
:GHS08
:GHS06
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520