1-Hexanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:2832-45-3
:MFCD00007542
Formula:C₆H₁₃O₃SNa
Melting Pt:300 °C
:M-Clarity™ quality level = MQ100
:188.22 g/mol
Storage Temperature:Ambient
EINECS:220-601-3
Specification Test Results
- Hill Formula:C₆H₁₃NaO₃S
- Molar Mass:188.22 g/mol
- pH value:5.5 - 7.5 (100 g/l, H₂O, 20 °C)
- Storage class:10 - 13 Other liquids and solids
- WGK:WGK 1 slightly water endangering
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Pharma Segment Brochure
Product Literature
msds_118305
SDS
ProductPage_118305
Product specification
Frequently Bought Together