ContactUsLinkComponent
1-Heptanesulphonic acid sodium salt ≥98.0% (by acidimetry) for HPLC, for ion pair chromatography, J.T.Baker®
  2173-05
 :  Avantor
 :  
undefined
1-Heptanesulphonic acid sodium salt ≥98.0% (by acidimetry) for HPLC, for ion pair chromatography, J.T.Baker®
  2173-05
 :  Avantor
 :  
  • Pack type:
    Glass bottle, amber, wide mouth
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:
  • For Laboratory,Research,or Manufacturing Use:
    ≥98.0%
  • Assay (as C₇H₁₅SO₃Na) (by Acidimetry):
    ≥98.0%
  • UV Absorbance of 0.25 M Solution - 250 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 240 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 230 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 220 nm:
    ≤0.060
  • UV Absorbance of 0.25 M Solution - 210 nm:
    ≤0.080
  • UV Absorbance of 0.25 M Solution - 200 nm:
    ≤0.200
 : Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate
 :
 :202.25 g/mol
 :245-210-5
 :GHS07
 :Ambient
 :22767-50-6
 :H₃C(CH₂)₅CH₂SO₃Na
 :MFCD00007543
 :300