

Ethyl methanesulphonate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:62-50-0
:
Formula:C₃H₈O₃S
MDL Number:MFCD00007559
:124.16 g/mol
Merck Index:13,03859
Boiling Pt:213 °C (1013 hPa)
Density:1.2079 g/cm³ (20 °C)
Flash Pt:100 °C
UN:2810
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:200-536-7
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:50 - 100 g/l (27 °C)
- Molar Mass:124.16 g/mol
- Boiling Point:213 - 214 °C (1013 hPa)
- Vapor Pressure:0.27 hPa (25 °C)
- Flash Point:100 °C
- Refractive Index:1.418 (20 °C, 589 nm)
- Density:1.206 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together