VWR® Basic, Frocks
Supplier: VWR Collection
About this item
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Press stud fastening
- Elastic wrists
- Raglan sleeves
- Free from natural rubber latex
Weight: 40 g/m²
: 1 piece per bag, 50 bags per case.
4 Options Available Below
Product Details & Documents
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Press stud fastening
- Elastic wrists
- Raglan sleeves
- Free from natural rubber latex
Weight: 40 g/m²
: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- 113-8995
- 113-8996
- 113-8998
- 113-8999
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- General Purpose|Laboratory
- General Purpose|Laboratory
- General Purpose|Laboratory
- General Purpose|Laboratory
- Weight:
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
Specifications
Product Family Options
Product Information
- ColourClothing SizeLengthAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-8995Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationGeneral Purpose|LaboratoryWeight40 g/m² - Specifications:
More
Specifications
Cat. No.113-8996Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationGeneral Purpose|LaboratoryWeight40 g/m² - Specifications:
More
Specifications
Cat. No.113-8998Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationGeneral Purpose|LaboratoryWeight40 g/m² - Specifications:
More
Specifications
Cat. No.113-8999Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationGeneral Purpose|LaboratoryWeight40 g/m²