Dithiothreitol (DTT, Cleland's reagent), Sigma-Aldrich®
ROCH10197777001
: ROCHE
:
- Pack type:Plastic bottle for solids
- Pk:2 g
- Seq:3107
:
:154.25 g/mol
:∼125 °C (1013 hPa)
:42…44 °C
:M-Clarity™ quality level = MQ100
:>110 °C
:Refrigerator
:222-468-7
:GHS07
:3483-12-3
:27565-41-9
:C₄H₁₀O₂S₂
:MFCD00004877