
Dithiothreitol (DTT, Cleland's reagent) for biochemistry, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
:154.25 g/mol
Boiling Pt:∼125 °C (1013 hPa)
Melting Pt:42…44 °C
:M-Clarity™ quality level = MQ200
Flash Pt:>110 °C
EINECS:222-468-7
:GHS07
Storage Temperature:Refrigerator
Related Information
Certificates
Search by Lot NumberMERC1.11474.0005_TDS_EN ProductPage_111474
Product specification
msds_111474
SDS
Frequently Bought Together