Order Entry
Austria
ContactUsLinkComponent
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure Bioreagent, J.T.Baker®
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure Bioreagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure Bioreagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Keep refrigerated between 2-8°C (36-46°F) and store under nitrogen.:
    = 0.060
  • Assay (HSCH₂(CHOH)₂CH₂SH):
    ≥99.5 %
  • Absorbance of a 0.1M Solution (1-cm path vs water) at - 280 nm:
    ≤0.060
  • Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm:
    ≤0.400
  • Appearance:
    Passes Test
  • Melting Point:
    40 - 43 °C

Specification Test Results

Related Information

Frequently Bought Together