
Dithiothreitol (DTT, Cleland's reagent) ≥99.4% for biotechnology
Catalog # 0281-25G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Abs.@283 nm (0.02 M, Water):≤0.05
- DNase:none detected
- Loss on Drying:<0.5%
- Melting Point:39 – 43 °C
- Oxidised DTT:<0.5%
- Protease:none detected
- Purity (-SH Content):>99.4%
- RNase:none detected
- Solubility (5%, Water):pass
Specification Test Results
About this item
:
:154.25 g/mol
Boiling Pt:< 125 °C (1013 hPa)
Melting Pt:42…44 °C
Flash Pt:>110 °C (open cup)
Storage Temperature:Refrigerator
EINECS:222-468-7
:GHS07
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877