


Methanol ≥99.9%, PESTINORM® for purge and trap GC analysis
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:11 °C (closed cup)
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Acidity:Max. 0.0005 meq/g
- Water:Max. 0.1 %
- Evaporation residue (100°C):Max. 0.0005 %
- 2-Butanone (GC/MS):Max. 0.5 ppm
- Volatile impurities (GC/MS):Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC83967.290 spec_VWRC83967.290 spec_VWRC83967.290.pdf spec_VWRC83967.320 spec_VWRC83967.320
Product specification
VWR® for chromatography columns, consumables and reagents
Product Literature
Frequently Bought Together