Order Entry
Austria
ContactUsLinkComponent
Ethanol 96% (v/v), SPECTRONORM® for spectroscopy
Flammable
Irritant
Ethanol 96% (v/v), SPECTRONORM® for spectroscopy
Catalog # 20822.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Ethanol 96% (v/v), SPECTRONORM® for spectroscopy
Catalog # 20822.290
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Assay (V/V):
    96,2-96,5%
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    5,2-7,5%
  • Transmittance (210 nm):
    Min. 35 %
  • Transmittance (220 nm):
    Min. 55 %
  • Transmittance (230 nm):
    Min. 72 %
  • Transmittance (240 nm):
    Min. 90 %
  • Transmittance (250 nm):
    Min. 95 %
  • Transmittance (from 270 nm):
    Min. 98 %

Specification Test Results

About this item

 :
 :46.07 g/mol
Boiling Pt:78.3 °C (1013 hPa)
Melting Pt:–117 °C
Density:0.7895 g/cm³ (20 °C)
Flash Pt:12 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119457610-43
 :GHS02
 :GHS07
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568