Order Entry
Austria
ContactUsLinkComponent
2-Propanol ≥99.5%, Laboratory Reagent
Flammable
Irritant
2-Propanol ≥99.5%, Laboratory Reagent
Certificates
undefined
Flammable
Irritant
2-Propanol ≥99.5%, Laboratory Reagent

Specification Test Results

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.5 %
  • Non-volatile matter:
    Max. 0.003 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Autoignition Temperature:
    750 °F
  • Vapor Density:
    2.1 (vs air)
  • Vapor Pressure:
    33 mmHg ( 20 °C)

Specification Test Results

Related Information

Frequently Bought Together