Propionic anhydride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-62-6
:
Formula:C₆H₁₀O₃
MDL Number:MFCD00009303
:130.14 g/mol
Merck Index:14,7826
Boiling Pt:166…168 °C
Melting Pt:–45 °C
Density:1.012
Flash Pt:63 °C (145 °F)
Storage Temperature:Ambient
EINECS:204-638-2
UN:2496
ADR:8,III
:GHS05
Related Information
Frequently Bought Together