ContactUsLinkComponent
Acetic anhydride, Sigma-Aldrich®
Acetic anhydride, Sigma-Aldrich®
  SIAL320102-1L
 :  Merck
 :  
undefined
Acetic anhydride, Sigma-Aldrich®
  SIAL320102-1L
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
 : Acetyl oxide, Acetyl acetate
 :
 :102.09 g/mol
 :203-564-8
 :1715
 :01-2119486470-36
 :GHS02
 :GHS05
 :GHS07
 :Ambient
 :108-24-7
 :(CH₃CO)₂O
 :MFCD00008705
 :139.9
 :–73 °C
 :1.08 g/cm³ (20 °C)
 :49
 :8 II
 :13,00057
 :M-Clarity™ quality level = MQ200