
Butyric anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-31-0
:
Formula:C₈H₁₄O₃
MDL Number:MFCD00009389
:158.2 g/mol
Merck Index:13,01594
Boiling Pt:198 °C (1013 hPa)
Melting Pt:–73 °C
Density:0.963 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-383-4
UN:2739
ADR:8,III
:M-Clarity™ quality level = MQ200
Flash Pt:83 °C
:GHS05
Specification Test Results
- Solubility:(20 °C) (rigorous decomposition)
- Melting Point:-75 °C
- Molar Mass:158.19 g/mol
- Boiling Point:195 - 199 °C (1013 hPa)
- Vapor Pressure:0.4 hPa (20 °C)
- Flash Point:87 °C
- Refractive Index:1.413 (20 °C, 589 nm)
- Explosion Limit:1.1 - 7.6 %(V)
- Density:0.963 g/cm3 (20 °C)
- Ignition Point:290 °C
Specification Test Results
Frequently Bought Together