Order Entry
Austria
ContactUsLinkComponent
 
Bis(2-methoxyethyl)ether ≥99% stabilised
Catalog # A13397.AP
CAS Number:  
Bis(2-methoxyethyl)ether ≥99% stabilised
Catalog # A13397.AP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

CAS Number:111-96-6
 :
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503
Merck Index:14,3165
Boiling Pt:161…162 °C
Melting Pt:–64 °C
Density:0.944
Flash Pt:51 °C (124 °F)
EINECS:203-924-4
UN:3271
ADR:3,III
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
 :134.18 g/mol
 :GHS08
 :GHS02
Storage Temperature:Ambient