
2-Butoxyethanol
Supplier: Roth Carl
Certificates
About this item
CAS Number:111-76-2
:
Formula:CH₃(CH₂)₃OCH₂CH₂OH
MDL Number:MFCD00002884
:118.18 g/mol
Merck Index:13,01558
Boiling Pt:171 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:61 °C
Storage Temperature:Ambient
EINECS:203-905-0
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together