Order Entry
Austria
ContactUsLinkComponent
2-Butoxyethanol ≥99%, TECHNICAL
Irritant
2-Butoxyethanol ≥99%, TECHNICAL
Catalog # 22122.363
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
2-Butoxyethanol ≥99%, TECHNICAL
Catalog # 22122.363
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

About this item

 :
 :118.18 g/mol
Merck Index:13,01558
Boiling Pt:171 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.902 g/cm³ (20 °C)
Flash Pt:50 °C
Storage Temperature:Ambient
EINECS:203-905-0
UN:2810
ADR:6.1,III
REACH:01-2119475108-36
 :GHS07
CAS Number:111-76-2
Formula:CH₃(CH₂)₃OCH₂CH₂OH
MDL Number:MFCD00002884