Order Entry
Austria
ContactUsLinkComponent
2-Butoxyethanol for synthesis, Sigma-Aldrich®
Irritant
2-Butoxyethanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Irritant
2-Butoxyethanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    118.17 g/mol
  • Boiling Point:
    168 - 170 °C
  • Vapor Pressure:
    0.8 hPa (20 °C)
  • Flash Point:
    67 °C
  • Refractive Index:
    1.4193 (20 °C, 589 nm)
  • Explosion Limit:
    1.1 - 10.6 %(V)
  • Density:
    0.90 g/cm3 (20 °C)
  • pH:
    7 (H2O, 20 °C) (as aqueous solution)
  • Ignition Point:
    230 °C
  • Saturation concentration:
    5 g/m3 (20 °C) Air
  • Viscosity kinematic:
    3.642 mm2/s (20 °C)
  • Evaporation number:
    160

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together