Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Butoxyethanol
2-Butoxyethanol ≥99%
2-Butoxyethanol ≥99%
:
Thermo Fisher Scientific
2-Butoxyethanol ≥99%
:
Thermo Fisher Scientific
:
111-76-2
:
:
CH₃(CH₂)₃OCH₂CH₂OH
:
MFCD00002884
:
118.18 g/mol
:
14,1559
:
171…173 °C
:
–79 °C
:
0.902
:
64 °C (147 °F)
:
Ambient
:
203-905-0
:
2810
:
6.1,III
:
GHS07
ALFAA17976.AP_TDS_EN
msds_ALFAA17976
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Ethanol absolute ≥99.9% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
: Merck
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
: Merck
Acetone, LiChrosolv® for HPLC, Supelco®
: Merck
Glycerine 85% Ph. Eur.
: VWR Chemicals
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
: VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals