Order Entry
Austria
ContactUsLinkComponent
Dibutyl ether ≥99.5% (by GC), Puriss. p.a. analytical reagent low in aromatic hydrocarbons, Riedel-de Haën™
Dibutyl ether ≥99.5% (by GC), Puriss. p.a. analytical reagent low in aromatic hydrocarbons, Riedel-de Haën™
Catalog # 34462-1L
CAS Number:  
Dibutyl ether ≥99.5% (by GC), Puriss. p.a. analytical reagent low in aromatic hydrocarbons, Riedel-de Haën™
Catalog # 34462-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Autoignition Temperature:
    365 °F
  • Assay (GC):
    Min. 99.5 %
  • Refractive index (n 20/D):
    1.398 - 1.400
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.1 ppm
  • Peroxides (as H₂O₂):
    Max. 20 ppm
  • Aromatic compounds (as Xylene):
    Max. 0.05 %
  • Refractive Index:
    n20/D 1.399 (lit.)

Specification Test Results

About this item

Synonyms: Di-n-butyl ether
 :
 :130.23 g/mol
Merck Index:13,01568
Boiling Pt:140 °C
Melting Pt:-95 °C
Melting Pt:–95 °C
Density:0.77 g/cm³
Flash Pt:8.5 %(V)
Storage Temperature:Ambient
EINECS:205-575-3
UN:1149
ADR:3,III
 :GHS02
 :GHS07
CAS Number:142-96-1
Formula:C₈H₁₈O
MDL Number:MFCD00009461