Dibutyl ether ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Di-n-butyl ether
CAS Number:142-96-1
:
Formula:C₈H₁₈O
MDL Number:MFCD00009461
:130.23 g/mol
Merck Index:14,1569
Boiling Pt:141…143 °C
Melting Pt:–95 °C
Density:0.764
Flash Pt:25 °C (77 °F)
Storage Temperature:Ambient
EINECS:205-575-3
UN:1149
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together