Dibutyl ether 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Di-n-butyl ether
CAS Number:142-96-1
:
Formula:C₈H₁₈O
MDL Number:MFCD00009461
:130.23 g/mol
Merck Index:13,01568
Boiling Pt:141 °C (760 mmHg)
Melting Pt:–95 °C
Density:0.768 g/cm³
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:205-575-3
UN:1149
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3980 to 1.4000 (20°C, 589 nm)
- Water:≤0.05 %
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.01 %
- Peroxides:≤0.01 % dibutylperoxide
Specification Test Results
Frequently Bought Together