VWR® Basic ESD Visitor Coats, SPP
113-8954
- Clothing Size:XXL
- Closure type:Press studs / snaps
- Garment application:Laboratory,Industrial
- Gender:Unisex
- Pockets:Without pockets
- Colour:White
- Length:112 cm
- Pk:50
- Weight:30 g/m²
Disposable coats, with collar, made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Press stud closure
- Elastic wrists
- Free from natural rubber latex
- Anti-static properties
Weight: 30 g/m²
: 1 piece per bag, 50 bags per case.