Order Entry
Austria
Orders LinkContactUsLinkComponent
Diethanolamine

Diethanolamine

Corrosive
Health
Irritant
Marketsource
Supplier:   Roth Carl
Diethanolamine
Supplier:  Roth Carl
Marketsource
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol
  • CAS Number:111-42-2
  • Formula:C₄H₁₁NO₂
  • MDL Number:MFCD00002843
  • Molecular Weight:105.14 g/mol
  • Merck Index:13,03134
  • Boiling Pt:268.8 °C (1013 hPa)
  • Melting Pt:28 °C
  • Density:1.1 g/cm³ (20 °C)
  • Flash Pt:176 °C
  • Storage Temperature:Ambient
  • EINECS:203-868-0
  • REACH:01-2119488930-28
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 48,70 €
    48,70 €
    1 * 250 mL
       48,70 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 86,70 €
    86,70 €
    1 * 500 mL
       86,70 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 141,00 €
    141,00 €
    1 * 1 L
       141,00 €

    Specifications:
    Marketsource
    Marketsource