Spermidine
Glass bottle for solids, 2,5 g
Catalog # ROTH7161.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
- CAS Number:124-20-9
- Formula:C₇H₁₉N₃
- MDL Number:MFCD00008229
- Molecular Weight:145.25 g/mol
- Merck Index:12,08893
- Boiling Pt:246 °C (1013 hPa)
- Melting Pt:20…25 °C
- Density:0.925 g/cm³ (20 °C)
- EINECS:204-689-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 g



