Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Dibutylamine for synthesis, Sigma-Aldrich®
 :  Merck
 
undefined
 
 
 
Dibutylamine for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    4.05 g/l (25 °C)
  • Melting Point:
    -62 °C
  • Molar Mass:
    129.24 g/mol
  • Boiling Point:
    160 °C (1013 hPa)
  • Vapor Pressure:
    2.3 hPa (20 °C)
  • Flash Point:
    39 °C DIN 51755 Part 1
  • Refractive Index:
    1.4168 (20 °C, 589 nm)
  • Explosion Limit:
    0.6 - 6.8 %(V)
  • Density:
    0.76 g/cm3 (20 °C)
  • pH:
    11.1 (1 g/l, H2O, 20 °C)
  • Ignition Point:
    260 °C DIN 51794
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

 

Frequently Bought Together