1,5-Diaminopentane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:462-94-2
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00008239
:102.18 g/mol
Merck Index:12,01645
Boiling Pt:178 °C (1013 hPa)
Density:0.9174 g/cm³ (0 °C)
EINECS:207-329-0
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:2735
:GHS05
Storage Temperature:Ambient
Frequently Bought Together