1,5-Diaminopentane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:462-94-2
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00008239
:102.18 g/mol
Merck Index:14,1609
Boiling Pt:178…180 °C
Melting Pt:9 °C
Density:0.873
Storage Temperature:Ambient
EINECS:207-329-0
UN:2735
ADR:8,II
:GHS05
Related Information
Frequently Bought Together