Order Entry
Austria
ContactUsLinkComponent
1,3-Diaminopropane for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Health
1,3-Diaminopropane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
Corrosive
Health
1,3-Diaminopropane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -12 °C
  • Molar Mass:
    74.12 g/mol
  • Boiling Point:
    137 - 140 °C (1013 mbar)
  • Vapor Pressure:
     - 10 hPa (20 °C)
  • Flash Point:
    50 °C
  • Refractive Index:
    1.4575 (20 °C, 589 nm)
  • Explosion Limit:
    2.8 - 15.2 %(V)
  • Density:
    0.89 g/cm3 (20 °C)
  • pH:
    12 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    350 °C

Specification Test Results

Related Information

Frequently Bought Together