Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®
: Merck
:77-86-1
:
:NH₂C(CH₂OH)₃
:MFCD00004679
:121.14 g/mol
:13,09842
:357 °C (1013 hPa)
:169…172 °C
:1.328 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:201-064-4
:GHS07
:Ambient
Frequently Bought Together