Order Entry
Austria
ContactUsLinkComponent
 
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, GPR RECTAPUR®
 
undefined
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 99 %
  • Melting point:
    169 - 172 °C
  • Ignition residue (SO4):
    Max. 0.2 %

 

Frequently Bought Together