Order Entry
Austria
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Catalog # MOLE26645271-1KG
Supplier:  Molekula
CAS Number:  
undefined
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Catalog # MOLE26645271-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
 :GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679