Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), EMPROVE® ESSENTIAL Ph. Eur., BP, JPC, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
Related Information
Certificates
Search by Lot NumberMERP1.08386.1000_TDS_EN MERP1.08386.5000_TDS_EN MERP1.08386.9025_TDS_EN ProductPage_108386
Product specification
msds_108386
SDS
Frequently Bought Together