
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Catalog # 17-1321-01
Supplier: Cytiva
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Application:For preparing electrophoresis buffer. Used in PAGE and Sequencing.
- Purity:≥99,8%
- pH Range:7.2 - 9.0
- Boiling/Condensation Point:219 - 220°C
- Melting/Freezing Point:170°C
- Solubility:Soluble in cold water and hot water, methanol
- Physical State:Solid
- Color:White
- Storage conditions:10 - 30°C
- Estimated shelf life from manufacture date:12 month
- Wetted Part:No
- Guidance:Tris = (tris(hydroximethyl)aminoethane)
- DIGE approved:Y
Specification Test Results
About this item
:
:121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
:GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679