Order Entry
Austria
ContactUsLinkComponent
 
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.8%, Proteomics Grade
Catalog # M151-1KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.8%, Proteomics Grade
Catalog # M151-1KG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Abs.@280 nm (1M, Water):
    0.05
  • Arsenic:
    <0.0005%
  • Copper:
    <0.0001%
  • DNase:
    NONE
  • Insolubles:
    0.005 %
  • Iron:
    <0.0001%
  • Lead:
    <0.0001%
  • Loss on Drying:
    1.0%
  • Magnesium:
    0.0005%
  • Protease:
    NONE
  • Purity:
    99.8%
  • RNase:
    NONE
  • Solubility (10%, Water):
    PASS

Specification Test Results

About this item

 :
 :121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
 :GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679