Order Entry
Austria
ContactUsLinkComponent
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Flammable
Corrosive
Irritant
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Catalog # PIER17919
CAS Number:  
undefined
Flammable
Corrosive
Irritant
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Catalog # PIER17919
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

About this item

 :
 :116.21 g/mol
Boiling Pt:120 °C (1013 hPa)
Melting Pt:–64 °C
Density:0.7765 g/cm³ (20 °C)
Flash Pt:17 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-744-6
UN:2372
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
CAS Number:110-18-9
Formula:C₆H₁₆N₂
MDL Number:MFCD00008335