Order Entry
Austria
ContactUsLinkComponent
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99% for biochemistry, for electrophoresis
Irritant
Corrosive
Flammable
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99% for biochemistry, for electrophoresis
Certificates
undefined
Irritant
Corrosive
Flammable
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99% for biochemistry, for electrophoresis

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4169 to 1.4189 (20°C, 589 nm)
  • Solubility:
    (10 % in water) Clear and no haze
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together