Triethylamine
: (Diethylamino)ethane, TEA
:121-44-8
:
:(C₂H₅)₃N
:MFCD00009051
:90
:–115 °C
:0.729 g/cm³ (20 °C)
:101.19 g/mol
:–11 °C
:Ambient
:1296
:3 II
:13,09740
:204-469-4
:GHS02
:GHS05
:GHS07
- Environmentally Preferable:
Frequently Bought Together