Order Entry
Austria
ContactUsLinkComponent
Iodoacetamide, Sigma-Aldrich®
Iodoacetamide, Sigma-Aldrich®
Catalog # SIALA3221-10VL
Supplier:  Merck
CAS Number:  
Iodoacetamide, Sigma-Aldrich®
Catalog # SIALA3221-10VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 Vial
  • Seq:
    0239

Specifications

About this item

 :
 :184.96 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:91…94 °C
Storage Temperature:Refrigerator
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:205-630-1
UN:2811
 :GHS07
CAS Number:144-48-9
Formula:ICH₂CONH₂
MDL Number:MFCD00008028