Iodoacetamide
Glass bottle for solids, 25 g
Catalog # MOLE24815520-25G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Iodoacetamide
- CAS Number:144-48-9
- Formula:ICH₂CONH₂
- MDL Number:MFCD00008028
- Molecular Weight:184.96 g/mol
- Boiling Pt:266 °C (1013 hPa)
- Melting Pt:91…94 °C
- Storage Temperature:Refrigerator
- EINECS:205-630-1
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



