Order Entry
Austria
ContactUsLinkComponent
Iodoacetamide, Millipore®
Irritant
Iodoacetamide, Millipore®
Catalog # 407710-25
CAS Number:  
Irritant
Iodoacetamide, Millipore®
Catalog # 407710-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :184.96 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:91…94 °C
Storage Temperature:Refrigerator
EINECS:205-630-1
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:144-48-9
Formula:ICH₂CONH₂
MDL Number:MFCD00008028