Order Entry
Austria
ContactUsLinkComponent
VWR® Advanced ESD Bouffant Caps
VWR® Advanced ESD Bouffant Caps
Catalog # 113-8901
Supplier:  VWR Collection
undefined
VWR® Advanced ESD Bouffant Caps
Catalog # 113-8901
Supplier:  VWR Collection

Specifications

  • Ø:
    580 mm
  • Colour:
    White
  • Pk:
    1.000

Specifications

About this item

These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.

  • Free from natural rubber latex
  • Anti-static properties

Weight: 16 g/m²

Packaging: 100 pieces per bag, 10 bags per case.